Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Colchicine ≥98%, Moligand™ cell culture reagent

Colchicine ≥98%, Moligand™ cell culture reagent

Health
Toxic
New Product
Supplier:   Aladdin Scientific
Colchicine ≥98%, Moligand™ cell culture reagent
Supplier:  Aladdin Scientific
New Product
Certificates
Colchicine ≥98%, Moligand™ cell culture reagent

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Colchicine, a tubulin Inhibitor, has been shown to block polymerization of microtubules by binding to tubulin. Studies indicate that this reagent also causes apoptosis of cells by acting on tissue plasminogen activators (tPA) and disrupting the cytoskeleton of cells. Exposure of cells to Colchicine has also been reported to result in alterations of Ca²⁺ concentrations in cells which may also result in apoptosis. A mitosis inhibitor useful in cell division studies.

Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-598-5
  • UN:1544
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $206.00
    $206.00
    Each(1/Itm/Bx)
       $206.00

    Supplier Number: C106740-1G
    Specifications:
    New Product
    New Product
  • Glass bottle
    500 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $124.00
    $124.00
    Each(1/Itm/Bx)
       $124.00

    Supplier Number: C106740-500MG
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.