Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bismuth(III) hydroxide ≥90%, Analytical reagent (AR)

Bismuth(III) hydroxide ≥90%, Analytical reagent (AR)

Irritant
Supplier:   Aladdin Scientific
Bismuth(III) hydroxide ≥90%, Analytical reagent (AR)
Supplier:  Aladdin Scientific
Certificates
Bismuth(III) hydroxide ≥90%, Analytical reagent (AR)

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hydrolysis of ribonucleic acid, absorbent. Used in detecting reducing sugars.

Synonyms:  Bismuth hydroxide
  • CAS Number:10361-43-0
  • Formula:H₃BiO₃
  • Molecular Weight:260.02 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $32.90
    $32.90
    Each(1/Itm/Bx)
       $32.90

    Supplier Number: B106150-100G
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $12.90
    $12.90
    Each(1/Itm/Bx)
       $12.90

    Supplier Number: B106150-25G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.