Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Agar

Agar

Plastic bottle, 500 g
Catalog # ALA109142-500G
Supplier:   Aladdin Scientific
Supplier #:  A109142-500G
Agar
Catalog # ALA109142-500G
Supplier:  Aladdin Scientific
CAS Number:  9002-18-0
Agar

Plastic bottle, 500 g

CAS Number:  9002-18-0
Price
$80.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Agar, often referred to as agar-agar is used as a solid substrate for inclusion in culture media, gel preparation, and microbiological work. With the addition of nutritional or selection supplements, it is often used as a growth medium for bacteria and fungi. Agar is an excellent matrix for work with proteins and nucleic acids, being ideally suited for the separation of large molecules. Widely used for immunodiffusion and chromatography applications. Porous, agarose gel in varying concentrations is used as a medium for assessing the motility and mobility of microorganisms; and may also be used in the evalution of chemotaxis and chemokinesis with regard to a cell population’s movement toward a chemoattractant.

Application: Agar has been used:as a component of the fruit fly dietas a component of saline-agar for the approximation of gray matter from brainin in vitro root nodulation assay of Glycine max seeds Agar has been used:in the preparation of laboratory diet to maintain the culture of D. melanogaster wild-type larvaas a high gel strength agar in the seed culture media to grow Arabidopsis thaliana mutant seedsto construct a tissue-mimicking phantom to test the imaging performance of an ultrasound transducer.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:C₁₂nH₁₈nOn₉n
  • MDL Number:MFCD00081288
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specification Test Results

  • Ash::
    ≤5.0%
  • High gel strength: :
    1000 - 1200 g/cm²

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.