Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tissue Optical Clearing Reagent, SCALEVIEW-A2

Tissue Optical Clearing Reagent, SCALEVIEW-A2

Optical tissue clearing reagent, 500 ml
Catalog # 77974-616
New Product
Supplier #:  193-18455
Tissue Optical Clearing Reagent, SCALEVIEW-A2
Catalog # 77974-616
New Product
CAS Number:  
Tissue Optical Clearing Reagent, SCALEVIEW-A2

Optical tissue clearing reagent, 500 ml

CAS Number:  
Price
$134.84
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

SCALEVIEW-A2 is reagent used for tissue clearing by ScaleA2 method.

  • Simple operation
  • Compatible with immunofluorescence

SCALEVIEW-A2 is reagent used for tissue clearing by ScaleA2 method. And the reagent used for antibody staining (AbScale method) and fluorescent staining (ChemScale method) of transparent tissue combining with SCALEVIEW-S series. Their methods were developed by Dr. Atsushi Miyawaki etc.

Fixed mouse and rat brain is made transparent only soaking into the reagent. Also, the reagent used for immersion liquid and to enable microscopic observation. 2 mm section can be made transparent in 2 weeks using the reagents.

Caution: For research use only.

Specifications

Specifications

  • Description:
    Optical tissue clearing reagent
  • Size:
    500 ml
  • Storage Conditions:
    Cool place (stored below 25 °C)
  • Cat. No.:
    77974-616
  • Form:
    Liquid
  • Shelf Life:
    24 months from the date of manufacture

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.