Orders LinkContactUsLinkComponent
Thioflavin T cell permeable ≥97% Fluorescent amyloid binding benzothiazole salt
500 mg
  ABCA_AB120751-500M
New Product
 :   Abcam
 :  AB120751-500MG
Thioflavin T cell permeable ≥97% Fluorescent amyloid binding benzothiazole salt
  ABCA_AB120751-500M
 :  Abcam
New Product
 :  2390-54-7
Thioflavin T cell permeable ≥97% Fluorescent amyloid binding benzothiazole salt
500 mg
 :  2390-54-7
$79.10
Each (500mg)
Fluorescent cell-permeable amyloid binding benzothiazole salt. Dye used to visualise stacked β sheets in vitroand in vivo. Binding induces a shift in excitation maximum (385 to 450 nm) and emission maximum (445 to 482 nm). Highly soluble in water. Limited ability to cross the blood-brain barrier.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  •  :2390-54-7
  •  :318.9 g/mol
  • Constituents::
    100% Thioflavin
  • Size:
    500 mg