Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Caffeine ≥99% Adenosine antagonist

Caffeine ≥99% Adenosine antagonist

New Product
 :   Abcam
Caffeine ≥99% Adenosine antagonist
 :  Abcam
New Product
 
Caffeine ≥99% Adenosine antagonist

 

 

Psychostimulant. Adenosine A₁ and A₂A receptor antagonist. Inhibits cAMP phosphodiesterases. Activates ryanodine receptor and induces intracellular calcium release.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 :  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  •  :58-08-2
  •  :194.19 g/mol
 

 

  • Constituents::
    100% Caffeine
Product Family Options

 

  • Size
     
     
  • 1 g
       
       
    Each (1g)
    Each (1g) - $63.30
    $63.30
    Each (1g)
       $63.30
    Minimum order quantity is 1

     : AB120240-1G
     
    New Product
    New Product
  • 5 g
       
       
    Each (5g)
    Each (5g) - $106.00
    $106.00
    Each (5g)
       $106.00
    Minimum order quantity is 1

     : AB120240-5G
     
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.