Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
FSL-1 TLR2/6 agonist

FSL-1 TLR2/6 agonist

100 µg
Catalog # ABCA_AB144863-100U
New Product
Supplier:   Abcam
Supplier #:  AB144863-100UG
FSL-1 TLR2/6 agonist
Catalog # ABCA_AB144863-100U
Supplier:  Abcam
New Product
CAS Number:  322455-70-9
FSL-1 TLR2/6 agonist

100 µg

CAS Number:  322455-70-9
Price
$343.00
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Toll-like receptor 2/6 (TLR2/6) agonist. Anti-herpetic activity. Exhibits clathrin-dependent internalization. Induces TNF-α in macrophages. NF-κB activator. Active in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Fibroblast-stimulating Lipopeptide
  • CAS Number:322455-70-9
  • Molecular Weight:1666.16 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% FSL-1

Specifications

  • Size:
    100 µg

Specifications