Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tridecanoic acid ≥99% Phospholipase A2 substrate

Tridecanoic acid ≥99% Phospholipase A2 substrate

100 mg
Catalog # ABCA_AB143889-100M
New Product
Supplier:   Abcam
Supplier #:  AB143889-100MG
Tridecanoic acid ≥99% Phospholipase A2 substrate
Catalog # ABCA_AB143889-100M
Supplier:  Abcam
New Product
CAS Number:  638-53-9
Tridecanoic acid ≥99% Phospholipase A2 substrate

100 mg

CAS Number:  638-53-9
Price
$148.00
Log in for availability
Each (100mg)
Minimum order quantity is 1
Product Details & Documents

Phospholipase A2 substrate. 13-carbon saturated fatty acid. Product of anaerobic biodegradation of n-hexadecane.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Molecular Weight:214.34 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Tridecanoic acid

Specifications

  • Size:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.