Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥98% antioxidant

L(+)-Ascorbic acid ≥98% antioxidant

100 g
Catalog # ABCA_AB142830-100G
New Product
Supplier:   Abcam
Supplier #:  AB142830-100G
L(+)-Ascorbic acid ≥98% antioxidant
Catalog # ABCA_AB142830-100G
Supplier:  Abcam
New Product
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥98% antioxidant

100 g

CAS Number:  50-81-7
Price
$127.00
Log in for availability
Each (100g)
Minimum order quantity is 1
Product Details & Documents

L-(+)-Ascorbic acid, antioxidant. Achieve your results faster with highly validated, pure and trusted compounds.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Molecular Weight:176.12 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Ascorbic acid

Specifications

  • Size:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.