Rapamycin ≥98% mTORC1 complex inhibitor
About this item
Immunosuppressant and antifungal agent. Complexes with FKBP-12 and inhibits mTOR (mammalian target of rapamycin). In complex with its cellular receptor, the FK506-binding protein (FKBP12), rapamycin binds and inhibits the function of the mammalian target of rapamycin (mTOR).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:53123-88-9
- Molecular Weight:914.2 g/mol
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Immunosuppressant and antifungal agent. Complexes with FKBP-12 and inhibits mTOR (mammalian target of rapamycin). In complex with its cellular receptor, the FK506-binding protein (FKBP12), rapamycin binds and inhibits the function of the mammalian target of rapamycin (mTOR).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:53123-88-9
- Molecular Weight:914.2 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::98% Rapamycin
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: AB120224-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: AB120224-5MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



