Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Permeabilization Buffer (Flow Cytometry), Abcam

Permeabilization Buffer (Flow Cytometry), Abcam

Permeabilization Buffer (Flow Cytometry), 100 ml
Catalog # ABCA_AB314681-100M
New Product
Supplier:   Abcam
Supplier #:  AB314681-100ML
Permeabilization Buffer (Flow Cytometry), Abcam
Catalog # ABCA_AB314681-100M
Supplier:  Abcam
New Product
CAS Number:  
Permeabilization Buffer (Flow Cytometry), Abcam

Permeabilization Buffer (Flow Cytometry), 100 ml

CAS Number:  
Price
$206.00
Log in for availability
Each (100ml)
Minimum order quantity is 1
Product Details & Documents

Permeabilization Buffer (ab314681) is a ready to use permeabilization buffer for intracellular immunofluorescence staining for microscopy or flow cytometry.

  • pH: 7.4

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Permeabilization Buffer (Flow Cytometry)
  • Size:
    100 ml
  • Cat. No.:
    ABCA_AB314681-100M
  • Storage Temperature:
    Store at -20°C.

Specifications