Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cell Lysis Buffer, Abcam

Cell Lysis Buffer, Abcam

New Product
Supplier:   Abcam
Cell Lysis Buffer, Abcam
Supplier:  Abcam
New Product
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called 1067 Cell Lysis Buffer.

  • Constituents: 1% Alcohol Ethoxylate, 0.88% Sodium chloride, 0.32% Tris HCl

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCA_AB288311-100M
  • ABCA_AB288311-400M
  • Storage Temperature:
  • Store at +4°C.
  • Store at +4°C.

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • ABCA_AB288311-100M
    Cell Lysis Buffer
    Cell Lysis Buffer
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $137.00
    $137.00
    Each (100ml)
       $137.00
    Minimum order quantity is 1

    Supplier Number: AB288311-100ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB288311-100M
    Storage Temperature
    Store at +4°C.
    New Product
  • ABCA_AB288311-400M
    Cell Lysis Buffer
    Cell Lysis Buffer
    400 ml
    Log in for availability
    Log in for availability
    Each (400ml)
    Each (400ml) - $290.00
    $290.00
    Each (400ml)
       $290.00
    Minimum order quantity is 1

    Supplier Number: AB288311-400ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB288311-400M
    Storage Temperature
    Store at +4°C.
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.