Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Nuclear Extraction Kit, Abcam

Nuclear Extraction Kit, Abcam

Nuclear Extraction Kit, 100 Test
Catalog # ABCA_AB221978-100T
New Product
Supplier:   Abcam
Supplier #:  AB221978-100TEST
Nuclear Extraction Kit, Abcam
Catalog # ABCA_AB221978-100T
Supplier:  Abcam
New Product
CAS Number:  
Nuclear Extraction Kit, Abcam

Nuclear Extraction Kit, 100 Test

CAS Number:  
Price
$238.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This Nuclear Extraction Kit is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in our transcription factor assay kits.

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Nuclear Extraction Kit
  • Size:
    100 Test
  • Cat. No.:
    ABCA_AB221978-100T
  • Storage Temperature:
    Store at Multi.

Specifications