Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cell Lysis Buffer, Abcam

Cell Lysis Buffer, Abcam

Cell Lysis Buffer, 15 ml
Catalog # ABCA_AB152163-15ML
New Product
Supplier:   Abcam
Supplier #:  AB152163-15ML
Cell Lysis Buffer, Abcam
Catalog # ABCA_AB152163-15ML
Supplier:  Abcam
New Product
CAS Number:  
Cell Lysis Buffer, Abcam

Cell Lysis Buffer, 15 ml

CAS Number:  
Price
$148.00
Log in for availability
Each (15ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

ab152163 is a 10X Cell Lysis Buffer used to lyse cells under non-denaturing conditions.

  • pH: 7.5 Constituents: 10% Triton X-100, 8.8% Sodium chloride, 3.15% Tris HCl, 1.12% Sodium pyrophosphate decahydrate, 0.38% EGTA, 0.29% Tetrasodium ethylene diamine tetraacetate, 0.216% disodium;1,3-dihydroxypropan-2-yl phosphate;hydrate, 0.19% Sodium orthovanadate, 0.001% Leupeptin

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Cell Lysis Buffer
  • Size:
    15 ml
  • Cat. No.:
    ABCA_AB152163-15ML
  • Storage Temperature:
    Store at -20°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.