Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CTGF (Full) ELISA Kit Wako

CTGF (Full) ELISA Kit Wako

CTGF(Full) ELISA Kit Wako, 96 tests
Catalog # 77974-806
New Product
Supplier #:  290-84701
CTGF (Full) ELISA Kit Wako
Catalog # 77974-806
New Product
CAS Number:  
CTGF (Full) ELISA Kit Wako

CTGF(Full) ELISA Kit Wako, 96 tests

CAS Number:  
Price
$913.72
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

This kit quantifies CTGF (Full) in human serum/plasma. By using antibodies targeting Module 1 and Module 4 of CTGF, it allows measurement of total CTGF including cleaved forms.

  • When combined with the CTGF (Full+N) ELISA Kit (292-84901), specific quantification of N-terminal CTGF is possible - useful for IPF (Idiopathic pulmonary fibrosis) and fibrotic disease research for human serum/plasma(EDTA)

CTGF is a secretory protein of approximately 38 kDa produced by umbilical vein and vascular endothelial cells. CTGF is composed of 4 domains, designated Modules 1 through 4. Module 1 is the IGF-binding domain, Module 2 the vWC domain, Module 3 the TSP-1 domain, and Module 4 the CT domain, and different factors bind to each domain. CTGF is reportedly involved in the proliferation and differentiation of chondrocytes and in cell adhesion, as well as being a major factor in tissue fibrosis, and has been proposed as a candidate biomarker for various fibrotic disorders. Meanwhile, it has been reported that CTGF in blood contains an N-terminal region in which the C-terminal region has been cleaved, giving rise to the problem that the amount of N-terminal CTGF in blood cannot be measured because it is mixed with platelet-derived full-length CTGF at the time of blood sample collection1).

N-terminal CTGF has been proposed as a potential biomarker for pulmonary fibrosis2). This product is an ELISA kit for detection of CTGF (Full) using monoclonal antibodies that recognize CTGF Module 1 and Module 4. The amount of N-terminal CTGF in blood can be measured when this product is used in combination with a kit for detecting CTGF (Full) and CTGF (N-terminal region) (Code No. 292-84901 CTGF (Full + N-terminal region) ELISA Kit Wako)1). 1) Miyazaki, O. et al. : Ann. Clin. Biochem., 47, 205 (2010). 2) Kono, M. et al. : Clin. Chim. Acta, 412, 2211 (2011).

Specifications

Specifications

  • Description:
    CTGF(Full) ELISA Kit Wako
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    7.81 - 500 pM
  • Sample Volume:
    Serum: 5 µl; Plasma (EDTA): 10 µl
  • Host:
    Mouse
  • Time to Results:
    Approx. 2 h 50 min
  • Detection Method:
    Colorimetric (TMB substrate, measured at 450 nm with 620 nm reference)
  • Cat. No.:
    77974-806
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    7.81 pM
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    CTGF
  • Sample Type:
    Serum, plasma (EDTA)
  • Shelf Life:
    12 months from the date of manufacture
  • Storage Temperature:
    Refrigerated (at 2...10 °C storage)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.