Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Proteometer-CV MD Buffer

Proteometer-CV MD Buffer

Proteometer-CV MD Buffer Packets (4X)
Catalog # 78068-524
New Product
Supplier:   NOVILYTIC, LLC
Supplier #:  BUF-PCV-003
Proteometer-CV MD Buffer
Catalog # 78068-524
Supplier:  NOVILYTIC, LLC
New Product
CAS Number:  
Proteometer-CV MD Buffer
Proteometer-CV MD Buffer

Proteometer-CV MD Buffer Packets (4X)

CAS Number:  
Price
$716.80
Log in for availability
Each
Product Details & Documents

4 sealed buffer packets specifically formulated for use with Proteometer-CV kit.

  • Buffer packets available for reorder when the Proteometer-CV Reactor, MD Unit, and/or Proteometer-CV reagent are not required

Intended for use with the Proteometer-CV kit.

Ordering information: Reorder in sets of 4 as needed.

Packaging: Supplied in a custom box with instructions for use. Store at room temperature.

Specifications

Specifications

  • Description:
    Proteometer-CV MD Buffer Packets (4X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.