G-Biosciences Sterile Dulbecco's Phosphate Buffered Saline with Magnesium and Calcium
Dulbecco’s phosphate buffered saline with magnesium and calcium is used to provide buffer system to maintain cell culture in the physiological pH range (7-7.6).
Composition: 137 mM sodium chloride, 0.5 mM magnesium chloride, 8.1 mM disodium phosphate, 0.9 mM calcium chloride, 1.47 mM monopotassium phosphate and 2.7 mM potassium chloride, pH7.3±0.3
- Sterile filtered (0.05 µ filter)
- Endotoxin tested (endodoxin free, endotoxin levels ≤1 EU/ml)
- Suitable for cell culture
- Nuclease and protease free
- Available in 1, 10×1, 10 and 20 L sizes
- Storage temperature: 4 °C
It is used in irrigating, transporting, and washing cells, maintain cell viability for limited time. D-PBS with magnesium and calcium is also used for molecular biology applications. Manufactured at a ISO9001:2015 facility under GMP guidelines.
Frequently Bought Together