Hydrogen Peroxide Single Analyte Photometer Kit
Hydrogen peroxide single analyte photometer kit
: I-2016
The CHEMetrics Hydrogen Peroxide SAM (Single-Analyte Photometer) test kit.
- Range 0 to 6.00 ppm Method: Ferric thiocyanate
Kit comes in a plastic case and contains everything needed to perform 30 tests: Vacu-vials kit, SAM Photometer, four AAA batteries, screwdriver, and instructions.
- Description:Hydrogen peroxide single analyte photometer kit