ContactUsLinkComponent
Capillary Electrophoresis System with UV/LIF, BioPhase 8800
Capillary Electrophoresis System with UV/LIF, BioPhase 8800
Catalog # 77995-568
Supplier:  SCIEX
New Product
undefined
Capillary Electrophoresis System with UV/LIF, BioPhase 8800
Catalog # 77995-568
Supplier:  SCIEX
Supplier Number:  5312959
New Product
Specifications
  • Description:
    BioPhase 8800
Specifications
About this item
BioPhase 8800 capillary electrophoresis system designed for efficiency and ease-of-use operation for purity and heterogeneity analysis of biotherapeutics and other charged molecular species.
  • UV light source and detection optics
  • Integrated 488 nm laser source and detection (source/detection system is self-calibrating and are controlled via software)
  • Touchscreen display allowing selection of pre-developed methods/sequences and system operation (also enables ad hoc system operation)
  • Integrated networking capability allowing for system operation via SCIEX control software enabling method development, sequence creation, and data analysis
  • Removable/interchangeable UV filter module housing up to two filters (220 and 280 nm filters included)
  • Interchangeable LIF filter capability
The BioPhase 8800 system with UV/LIF includes the BioPhase 8800 instrument with integrated UV detection and integrated 488 nm laser for laser induced fluorescence. Includes liquid temperature stabilization module for capillary cartridge temperature control, sample and buffer plate temperature control, integrated touchscreen interface for at-instrument system monitoring and applications training. Compatible with SCIEX BioPhase cartridges, validated analysis kits, and software.
BioPhase 8800 system supply compatibility: BioPhase Sample, Reagent, and Outlet Plates designed to ANSI standard configuration; Pre-assembled 8-capillary cartridges (bare fused silica or neutral capillary); Validated assay kits: BioPhase CE-SDS Protein Analysis Kit, BioPhase Capillary Isoelectric Focusing (cIEF) Kit, RNA 9000 Purity and Integrity Kit.