About this item
The ImageXpress® HCS.ai High-Content Screening System captures your imagination with high-quality imaging and analysis, enabling you to acquire the data you need with ease.
Adapt to future lab needs with easy, cost-effective, on-site upgrades, protecting your initial investment
- Capture exceptional detail in 2D and 3D biological models with swappable spinning disks, optimized pinhole deep tissue disks, and advanced water immersion objectives
- Get clearer images faster with a highly sensitive 95% peak quantum efficiency sCMOS camera
- Streamline complex workflows with the upgraded MetaXpress® Acquire software, accessing robust and insightful data tailored to your needs
- Gain deeper insights with AI-powered IN Carta® Image Analysis Software and the ability to train your own deep learning models within minutes
- Go from image acquisition to data analysis all in a single platform
The system can be easily configured to meet your exact assay requirements, allowing you to perform a vast range of assays in a single solution. Benefit from up to 12 effective magnifications in a single system, and AgileOptix™ technology which offers a combination of a powerful solid-state light engine, custom optics, scientific CMOS sensor, and the ability to change between different disk geometries.
Empower your team to obtain high-definition images and robust data from a wide range of 2D and 3D cell assays, and leverage the power of AI for deeper insights.
Related Information
Evaluating Compound Toxicity Effects on Healthy Intestinal Organoids Using High-Content Imaging.pdf High-Speed Phenotypic Profiling Using a Next Generation High-Content Imaging Platform with AI-Enabled Image Analysis.pdf Using AI-Analysis Tools and a Next-Generation High-Content Screening Platform to Improve 3D Screening Assay Data Capturing.pdf