Parker ALIGN™ Multi-Gas Generator for LC-MS
Product Details & Documents
ALIGN™ Series of gas generators designed specifically for in-laboratory use, serving LC-MS/MS instruments. ALIGN™ generators are a total solution to providing the laboratory-grade gases required by mass spectrometers - plug-and-play systems that will perfectly meet the needs of the instrument.
Variable frequency drive saves up to 30% utility costs
Exceeds gas requirements for all SCIEX Instruments, including 7500 and Echo
Fits easily under all lab benches
Made in the USA
ALIGN™ Series generators are state-of-the-art machines featuring the best energy efficiency and gas purity on the market. Smart controllers enable the generators to operate only as much as the instrument calls for, thereby conserving energy, reducing electric bills, eliminating unnecessary noise, and increasing the reliability of the entire system. The ALIGN™ platform features a low-profile and small footprint designed to fit under the shortest lab bench, leaving benchtop space available for other valuable instruments.
ALIGN line of generators offer several key advantages over delivered gas cylinders or dewars. On-site gas generation eliminates the safety hazards created by moving and storing high-pressure cylinders and prevents any potential contamination from outside personnel. Generators also eliminate worry about your supply chain, any variation in gas quality from tank to tank, and keep the most valuable employees focused on the most profitable work.
Purity % Nitrogen: Up to 99%
Purity % Organics: 99.999%
Nitrogen (Curtain): 19 SLPM at 80 psi
Air 1 (Source): 25 SLPM at 110 psi
Air 2 (Exhaust): 25 SLPM at 110 psi
Dewpoint: −40 °F
Noise level: <55 dB(A)
Electrical: 230 VAC +/- 10%, 50/60 Hz, 1 Phase, 20 Amps, NEMA 6-20R Receptacle
Certifications: Stringent ISO 8573-1 Class 1.2.1 Compliant.
Specifications
- Description:ALIGNTM Multi-gas generator
- H×W×D:24.62×23.75×34.5”
- Voltage:230 VAC
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



