CentriVap Cold Traps are critical components designed to collect evaporated liquid from CentriVap Vacuum Concentrators. By condensing and trapping volatile solvents, they protect the vacuum pump and the laboratory environment from the harmful effects of vapors, fumes, and corrosives, while also ensuring that solvents can be easily collected for analysis or disposal.
230 V models, not for use in USA
For samples containing water
These traps are available either as integral components within configured CentriVap concentration systems or separately, making them suitable for a wide range of laboratory applications such as freeze drying and desiccation. Their role in maintaining system efficiency, safety, and solvent recovery makes them an essential part of many lab workflows.
230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (78007-958 and 78007-974) are also available, please see separate product page.