Stackable Drum Rack and Sumps, Galvanized Steel
Supplier: DENIOS INC
New Product
Certificates
About this item
Each rack is easily lifted and relocated with a forklift. The sumps catch drips and spills associated with dispensing, maintaining a neat, safe area.
All components are made of heavy gauge steel, galvanized to provide years of service with a rust-free appearance
- Sumps are 100% tested and leak-free guaranteed
- Constructed using premium 11-gauge quality steel
- Made from all- steel construction for an extended life
- Finished with a durable, hard-shell finish
- For secondary drum containment
- Ideal for oil, chemical, and universal secondary containment
- For 55 gallon drum containment or small cans
Spill containment sumps provide superior performance in containment, ensuring full compliance with industry standards for spill prevention. They are designed for long-term durability and effective environmental protection, making them an ideal choice for a wide range of applications.
The 1 Drum galvanized steel sump without a platform insert provides spill containment without the extra height and expense of forklift accessible base designs.
Certifications:
SPCC, OSHA, NFPA, UFC.
Frequently Bought Together