Order Entry
Puerto Rico
ContactUsLinkComponent
ESF AdvanCD™Chemically Defined Insect Cell Culture Medium, Expression Systems
ESF AdvanCD™Chemically Defined Insect Cell Culture Medium, Expression Systems
Catalog # 77705-736
New Product
CAS Number:  
ESF AdvanCD™Chemically Defined Insect Cell Culture Medium, Expression Systems
Catalog # 77705-736
Supplier Number:  54-018-01
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Bottle
  • Size:
    1 L
  • Shelf life:
    12 months from date of manufacture
  • Application:
    Insect cell types

Specifications

About this item

ESF AdvanCD™ is a 100% chemically defined, animal-origin-free, protein-free, serum-free insect cell culture medium that does not contain hydrolysates. This proprietary formula is manufactured exclusively by Expression Systems for exceptional performance while improving the consistency and efficiency of your process. ESF AdvanCD media was developed for robust cell growth in parallel with clonal Sf9 cell lines and also supports the growth of Trichoplusia cell lines.

  • Supports growth of suspension Sf9 cell lines in excess of 35 x 106 cells/ml

    Enables superior protein expression yields as compared to alternative media formulations

  • Demonstrates high reproducibility and lot-to-lot consistency

    Supports production of high titer baculovirus stocks with titers greater than 5×109 iu/ml

  • Maintains high baculovirus titers with reduced aggregation when coupled with VSA (Virus Stabilization Additive)
  • Supports scalable baculovirus infection and protein expression in Spodoptera and Trichoplusia cell lines
  • Adapted for use with a Sf9 rhabdovirus negative cell line for a fully compliant manufacturing platform
  • Ready-to-use liquid formulation, with no supplementation required