Order Entry
Puerto Rico
ContactUsLinkComponent
Hydrochloric acid Dilution Matrix 10 g/l Ph. Eur., Paragon Scientific
Hydrochloric acid Dilution Matrix 10 g/l Ph. Eur., Paragon Scientific
Catalog # 77975-752
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Hydrochloric acid Dilution Matrix 10 g/l Ph. Eur., Paragon Scientific
Catalog # 77975-752
Supplier:  LGC Standards
Supplier Number:  ALK-HCL10GL
New Product
CAS Number:  

Specifications

  • Size:
    1 l

Specifications

About this item

CAS Number:7647-01-0
 :
Formula:HCl
MDL Number:MFCD00011324
 :36.46 g/mol