Undercounter Refrigerators, TSU
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Thermo Scientific TSU series undercounter laboratory refrigerators offer a compelling cold storage option for a wide variety of laboratories. With a full suite of alarms, whisper quiet operation, and NSF 456 certified performance, the TSU series is designed specifically for laboratory settings.
Green refrigerants support global greenhouse gas emission reduction initiatives
Hi/Lo temperature, door ajar, and power fail alarms
Certifications:
cULus, ENERGYSTAR, NSF 456 certified.
Specifications
- Catalog No:
- 77995-562
- 77995-564
- Material:
- Painted steel
- Painted steel
- Compressor:
- Forced-air
- Forced-air