Immobilized Coenzyme A
Catalog # 77448-812
Supplier: G-Biosciences
CAS Number:
Specifications
- Size:1 g dry resin
Specifications
About this item
Immobilized Coenzyme A is made by covalently coupling Coenzyme A with 6% cross linked agarose beads. Cross linking is done at the amine on nucleotide with six-carbon space arm. Cross linking at the thiol group can be available for custom production.
Coenzyme A coupled to 6% cross-linked agarose
- Extent of labeling is 1.5 to 5 µmol coenzyme A per ml resin
- Shipped as lyophilzed dry resin
-
Available in 1 g size
Approximately 5 ml resin when hydrated
Application(s): Immobilized Coenzyme A is used in affinity purification of Coenzyme A binding enzymes.