ContactUsLinkComponent
Dry Block Heaters, Digital
Dry Block Heaters, Digital
Supplier:  Ohaus
New Product
Certificates
undefined
Dry Block Heaters, Digital
Supplier:  Ohaus
New Product
Specifications
  • Catalog No:
  • 77874-092
  • 77874-098
  • 77874-100
  • Model:
  • HB1DG
  • HB2DG
  • HB2DGHL
  • Temperature Stability:
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • Power:
  • 110 W (consumption)
  • 210 W (consumption)
  • 400 W (consumption)
Specifications