Diversey™ Wiwax ProX Maintenance Emulsion
Catalog # 77929-438
Supplier: Janitorial Supplies
New Product
Specifications
- Description:Wiwax ProX maintenance emulsion
- Packaging:Bottle
Specifications
About this item
An integral part of a comprehensive floor care program for cleaning, stripping, and sealing rubber floors.
Provides the floor with a shiny appearance
- Provides protection when used on a regular basis
Wiwax ProX Maintenance emulsion is a unique blend of waxes, polymers, and detergents to effectively rejuvenate and maintain rubber as well as linoleum, asphalt, marble, stone, concrete, and slate flooring.