Laboratory Ball Valve Lockout
77942-614
New Product
- Color:Red
- Description:Laboratory ball valve lockout
- W×D×H:3.125×1.125×3.5"
Laboratory ball valve lockouts secure lever-handle ball valves in lab settings.
Locks out laboratory ball valves with lever handles for enhanced safety
- Durable PETG construction for long-lasting use
- 1" diameter opening fits securely over the valve body
- Accommodates a single padlock (up to 5/16" shackle) for secure lockout
Laboratory Ball Valve Lockouts lock out ball valves with lever handles typically used in laboratory settings. The device is made from durable PETG plastic and has a 1" diameter opening that fits over the valve body. It accommodates a single padlock with up to a ⁵/₁₆" diameter shackle. Made in the USA.