Water ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®
EM1047351000
New Product
:
- Pack type:Borosilicate glass bottle
- Size:1 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- EPA 533/537.1 Tested:Conforms
- UHPLC Gradient (at 210 nm):≤ 2 mAU
- Evaporation Residue:≤ 0.7 mg/l
- UHPLC Gradient (at 254 nm):≤ 0.5 mAU
- (PEG) Signal-to-Noise Ratio:≤50
:7732-18-5
:
:H₂O
:MFCD00011332
:18,02 g/mol
:Room Temperature