ContactUsLinkComponent
DURAN® Original GL 25 Laboratory Bottle Screw Cap, with Lip Seal, PP, Blue
DURAN® Original GL 25 Laboratory Bottle Screw Cap, with Lip Seal, PP, Blue
Catalog # 77623-798
New Product
undefined
DURAN® Original GL 25 Laboratory Bottle Screw Cap, with Lip Seal, PP, Blue
Catalog # 77623-798
Supplier Number:  292391307
New Product
Specifications
  • Ø:
    33 mm
  • Color:
    Blue
  • Height:
    19 mm
  • Material:
    PP
Specifications
About this item
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colors: Blue, green, yellow and gray with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
  • Cap with integral lip seal provides effective sealing for short term storage
  • Pouring ring for drip-free pouring
  • The color helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
    Autoclavable
Fully compatible with the GL 45 threads and therefore highly versatile.