ContactUsLinkComponent
Eisco® Buzzer in Vacuum Jar Kit
Eisco® Buzzer in Vacuum Jar Kit
Catalog # 470343-978
Supplier:  Wards
undefined
Eisco® Buzzer in Vacuum Jar Kit
Catalog # 470343-978
Supplier:  Wards
Supplier Number:  PH0178
Specifications
  • Height:
    139.7 mm (5.5")
  • Diameter:
    100 mm (3.94")
Specifications
About this item
Airtight chamber reveals how sound disappears without air.
  • Built-in buzzer activates via external switch
  • Clear, impact-resistant acrylic construction
  • Compatible with standard hand vacuum systems
This apparatus helps students explore the fundamental role of air in sound transmission. Once sealed and evacuated, the chamber isolates the buzzer's vibrations from the surrounding environment, creating a silent visual paradox. Ideal for physics demonstrations, the setup encourages critical thinking and discussion. A vacuum pump is required for full functionality and must be sourced separately.