VWR® Fluorometer
About this item
VWR® Fluorometer for quantitation of DNA, RNA, and Proteins. Compact with touch screen control panel. Compatible with VWR® quantitation kits and kits from leading suppliers.
Sensitive quantitation down to 0.1 pg/µl ds DNA
- Large color touch screen for programming and operation
- Two models for single or eight-sample processing
- Built-in reagent calculators for sample preparation
- Compact and portable (11×6" footprint)
Sample volume: 1 to 20 µl
Compatible sample tubes: 0.5 ml transparent thin wall PCR tube
Linearity range: Five orders of magnitude
Sensitivity: dsDNA: 0.01 ng/µl, ssDNA: 0.05 ng/µl, RNA: 0.25 ng/µl, microRNA: 0.05 ng/µl
Calibration: 2 or 3 points
Control panel: 5" color touch screen
Working environment: 15 to 30 ℃, <75% RH, indoor use
Certifications: Meets ASTM standards. EN-342.
Accessories information: Accessories not included.
2 Options Available Below
Product Details & Documents
VWR® Fluorometer for quantitation of DNA, RNA, and Proteins. Compact with touch screen control panel. Compatible with VWR® quantitation kits and kits from leading suppliers.
Sensitive quantitation down to 0.1 pg/µl ds DNA
- Large color touch screen for programming and operation
- Two models for single or eight-sample processing
- Built-in reagent calculators for sample preparation
- Compact and portable (11×6" footprint)
Sample volume: 1 to 20 µl
Compatible sample tubes: 0.5 ml transparent thin wall PCR tube
Linearity range: Five orders of magnitude
Sensitivity: dsDNA: 0.01 ng/µl, ssDNA: 0.05 ng/µl, RNA: 0.25 ng/µl, microRNA: 0.05 ng/µl
Calibration: 2 or 3 points
Control panel: 5" color touch screen
Working environment: 15 to 30 ℃, <75% RH, indoor use
Certifications: Meets ASTM standards. EN-342.
Accessories information: Accessories not included.
Documents
VWR® Fluorometer Brochure.pdf
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- VWRI210-000173
- VWRI210-000157
- Light Source:
- LED
- LED
- Detector:
- Silicon photomultiplier
- Silicon photomultiplier
- Sample handling:
- 8 (sample throughput)
- 1 (sample throughput)
- Scanning speed:
- ≤5 s/sample
- ≤5 s/sample
- Electrical:
- 100 - 240 VAC
- 100 - 240 VAC
- W×D×H:
- 10.8×24.1×5.7 cm (4.25×9.5×2.25")
- 10.8×24.1×5.7 cm (4.25×9.5×2.25")
- Weight:
- 0.5 kg (1.1 lbs.)
- 0.5 kg (1.1 lbs.)
Specifications
Product Family Options
Product Information
- WavelengthAvailabilityPrice
- VWRI210-000173VWR® Fluorometer, 8-channel, 115 - 240 VVWR® Fluorometer, 8-channel, 115 - 240 VExcitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nmLog in for availabilityEach$7,237.40Specifications:New Product
More
Specifications
Cat. No.VWRI210-000173Light SourceLEDDetectorSilicon photomultiplierSample handling8 (sample throughput)Scanning speed≤5 s/sampleElectrical100 - 240 VACW×D×H10.8×24.1×5.7 cm (4.25×9.5×2.25")Weight0.5 kg (1.1 lbs.)New Product - VWRI210-000157VWR® Fluorometer, 100 - 240 VACVWR® Fluorometer, 100 - 240 VACExcitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nmLog in for availabilityEach$2,226.42Specifications:
More
Specifications
Cat. No.VWRI210-000157Light SourceLEDDetectorSilicon photomultiplierSample handling1 (sample throughput)Scanning speed≤5 s/sampleElectrical100 - 240 VACW×D×H10.8×24.1×5.7 cm (4.25×9.5×2.25")Weight0.5 kg (1.1 lbs.)
Accessories
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA HS quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× ready-to-use DS-DNA hs quant kit, 100 assaysVWR® 1× ready-to-use DS-DNA hs quant kit, 100 assaysVWRI210-000160Log in for availabilityEach - $135.20
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA BR quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× ready-to-use DS-DNA BR quant kit, 100 assaysVWR® 1× ready-to-use DS-DNA BR quant kit, 100 assaysVWRI210-000162Log in for availabilityEach - $135.20
Product Information
- Catalog #AvailabilityPrice
- VWR® SS-DNA quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA HS quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA BR quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® fluorometer tubes 0.5 ml, 250 per packVWR® fluorometer tubes 0.5 ml, 250 per packVWRI210-000166Log in for availabilityPack of 250 - $72.80
Product Information
- Catalog #AvailabilityPrice
- Fluorometer tubes, 0.2 ml, pk/1000
Product Information
- Catalog #AvailabilityPrice
- Fluorometer tube-strips, 0.2 ml, pk/120
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× DS-DNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA BR quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× DS-DNA BR quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® SS-DNA quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA BR quant kit, 500 assays



