The Sartolab® P20 Pump is a specialized fluid feed pump optimized for use with Sartolab® P20 pressure filtration devices. Sartorius recommends the Liquiport PML 16980 pump by KNF as the ideal companion for these filters, ensuring efficient sample processing. A key safety feature of this pump is the integrated pressure relief valve, which is pre-set to a maximum of 3.5 bar relative pressure. This valve serves as an essential protective mechanism, automatically activating to relieve pressure in the event of filter blockage that results in pressure buildup, thereby maintaining a secure and reliable operation.
Safety mechanism - Equipped with an integrated pressure relief valve for secure operation
- Compact design - Space-saving with a small footprint for convenient placement
- User-friendly interface - Simple controls for hassle-free operation
- Dry running capability - Self-priming pump that can operate without fluid
Robust construction:
Splashproof with an IP 65 rating, suitable for challenging environments
Brushless DC motor for less maintenance
Integrated overheating protection for longer lifetime
Versatile control:
Dual-mode functionality, offering both analog and impulse control (RC) capabilities
The RC option includes a dedicated interface for foot switch integration, significantly improving the ease and flexibility of operation
Applications:
Biotechnology and life sciences with Sartolab® P20 Devices
Chemical processing and synthetic organic chemistry
Pharmacological research and development
Production and testing of adhesives, glues, and coatings
Polymer research and polymer surface treatments
Food technology and research applications
Handling of detergents, solvents, and other chemical agents
- Catalog No:
- 77763-000
- 77762-998
- 77763-004
- 77763-002
- Inlet connection:
- Luer female connectors
- Max. Pressure:
- 3.5 bar rel
- Max. Temperature:
- Ambient: 5…40 °C; Liquid: 5…80 °C
- Outlet connection:
- Luer female connectors