Saturix Presaturated Cleanroom Wipes, 70% Presaturated Sachets
Catalog # 77871-042
Supplier: FG CLEAN WIPES TE
New Product
Specifications
- Material:Polyester/cellulose
- Environmentally Preferable:
- Packaging:Individually wrapped, sachet packets
- W×L:10.2×7.6 cm (4×3")
- Solvent:70% IPA, 30% DI water
Specifications
About this item
Convenience, consistency, and cleanliness.
Reduces IPA use and minimizes bulk liquid storage
- Reduces airborne VOC’s
- Provides consistency in solvent application
- Multiple sizes and packaging configurations in different formats and IPA concentrations
- Variety of substrates for different critical environments
- Easier and safer to ship, store, and use
This family of presaturated wipes feature wide-ranging substrates, IPA saturant levels, and packaging configurations for a multitude of environments and applications.
Packaging:
Wipes are packaged 100 per box, 10 boxes per case.