Order Entry
Puerto Rico
ContactUsLinkComponent
Polypropylene Bottle with Screw-On Cap
Polypropylene Bottle with Screw-On Cap
Catalog # BK356855
Supplier:  Beckman Coulter
Polypropylene Bottle with Screw-On Cap
Catalog # BK356855
Supplier:  Beckman Coulter
Supplier Number:  356855

Specifications

  • Capacity:
    750 ml
  • Description:
    Bottle with screw-on cap
  • O.D.×H:
    96×130 mm

Specifications

About this item

Bottles are supplied with screw-on caps for use in benchtop centrifuges and microcentrifuges rotors.

  • Polypropylene construction

    Screw-on closures included

  • Nominal capacity: 750 ml

Bottles are manufactured under ISO 9001 guidelines.