- Description:Adhesive tissue 3M
Vetbond is an n-butyl Cyanoacrylate adhesive that quickly and conveniently closes minor wounds. It cures in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occur.
Bonds tissue
Use in place of sutures for small animal surgery
Blue dye for easy-to-see drop application
Antimicrobial effect
Use in forensic science
Butyl Cyanoacrylate
Low toxicity
Cures in < 10 seconds
Compared with the traditional suture, vetbond adhesive has several advantages.
Close incisions in one-tenth of the time
Bonding strength is equal to 5-0 monofilament sutures
Antimicrobial effect that can decrease infection rates in contaminated wounds
Bonding sloughs off naturally in 5 to 7 days requiring no followup
Cosmetic appearance of the healed incision is also better