Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Carbon monoxide sensor
Specifications
About this item
Carbon monoxide (CO) is a versatile mediator of physiological processes. Carbon monoxide (CO) formed by internal mechanisms (endogenous) is measured in a variety of ways, but standard measurement methods are of limited utility in most biological systems. CO sensor measures CO in vivo or in vitro in real time.
Response time: <10 sec
Lowest detection limit: ~10 nM
Sensitivity: ~0.5 pA/nM
Application: Cell cultures
Measure CO in vivo or in vitro. Real time measurement and recording of dissolved CO. Works with WPI’s TBR free radial analyzers. 2.0 mm Satinless steel sensor with replaceable membrane sleeves.
This CO sensor is comprised of a 2.0 mm stainless steel body with a replaceable membrane-covered sleeve. The sleeve is filled with electrolyte. It is an amperometric sensor designed for use in cell culture and similar applications. CO diffuses through the gas-permeable membrane and is then oxidized to CO₂ on the working electrode of the sensor. This oxidation creates a current with a magnitude directly related to the concentration of CO in solution.
Package of 1.