About this item
ISO NOPF electrodes are flexible sensor, nearly unbreakable and available in 100, 200 and 500 µm diameters. Laboratory have created these completely flexible and virtually unbreakable NO sensors.
Excellent selectivity to NO
Rapid response time
Highly sensitive
Lowest detection limit: 0.2 nM
Choose diameter, length and shape
Poise voltage: 865 mV
The new sensors are based on a composite graphite NO-sensing element combined with a reference electrode. The surface of the sensor is then coated with a unique multi-layered NO-selective membrane.
These sensors are based on a composite graphite NO-sensing element combined with a reference electrode. The surface of the sensor is then coated with a unique multi-layered NO-selective membrane.
Requires no sleeves or filling solutions. Flexible and durable, like other ISO-NOPF sensors. High sensitivity for a rapid response time - ten times more sensitive than the ISO-NOP. Can be used in acidic conditions. Longer sensor tip than the ISO-NOP. Bigger linear range than the ISO-NOP (range is based on the length of the sensor tip). Calibration with either the SNAP or nitrite method.
It takes three days to test prior to shipping.
ISO NOPF200 package of 2; ISO NOPF100 package of 2; ISO NOPF500-CXX package of 2; ISO NOPF200-L10 package of 3.