Order Entry
Puerto Rico
ContactUsLinkComponent
Low-Lint Disposable Hand Drying Wipes
Low-Lint Disposable Hand Drying Wipes
Catalog # 77705-712
Supplier:  Contec
New Product
undefined
Low-Lint Disposable Hand Drying Wipes
Catalog # 77705-712
Supplier:  Contec
Supplier Number:  CRHW0001
New Product

Specifications

  • Color:
    White
  • Packaging:
    100/bag
  • W×L:
    43×23 cm (17×9")

Specifications

About this item

Absorbent, disposable hand and elbow drying wipes.

  • Protects against potential contaminants and gets stacks organized for ease in dispensing

  • Packaging minimizes the risk of airborne contamination
  • Wipe packaging can be easily wiped down with a disinfectant or sterile 70% IPA

Proper hand hygiene during garbing is critical to maintain compliance in cleanroom environments. Contec’s hand drying wipes are absorbent and disposable to prevent cross-contamination. These wipes are ideal for drying hands and forearms after hand washing.