Refrigerated Incubators
New Product
Thermo Scientific Refrigerated Incubators utilize robust compressor cooling to help ensure a controlled temperature environment to fit your application needs.
115 V models
Temperature range 4 to 60 °C
- Temperature stability and uniformity fitting most applications
- Environmentally-friendly hydrocarbon refringent, R290, has a Global Warming Potential index (GWP) of three, keeping the environmental impact of the refrigeration system minimized
- 7" touch screen for data export, alarm history, operating status, and real-time temperature curve
- Coated cabinet interior and adjustable shelves make cleaning easy, with flexible and corrosive-resistant operation
- USB data export
- Temperature sensor open, short cut-off alarm, door open alarm, standby or auto-restart after power failure
- High temperature cut-off and low temperature alarm
- Ability to program and store up-to 10 programs, with 99 segments each on user interface
- Three level access control to avoid non-authorized operation
Available in four sizes, these incubators are excellent for applications where results demand precisely controlled temperatures, including the development of new products, quality control in food, beverage and cosmetics industries, waste-water testing, cell culture, growth and preservation of samples, the equipment is always designed with your samples in mind.
:
CE, UL, CSA.
- Catalog No:
- 77889-372
- 77889-376
- 77889-380
- 77889-384
- Model:
- 77889-372
- 77889-376
- 77889-380
- 77889-384
- Shelves supplied (max.):
- 2 (5)
- 3 (7)
- 3 (7)
- 4 (9)