Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured SDA 200 Proof
Ethanol denatured SDA 200 Proof
Catalog # 10141-264
Supplier:  Decon Labs
CAS Number:  
Ethanol denatured SDA 200 Proof
Catalog # 10141-264
Supplier:  Decon Labs
Supplier Number:  2955M
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Size:
    55 Gal

Specifications

About this item

 :
 :46.07 g/mol
 :GHS02
 :GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568