Mouse Recombinant EBI3 Subunit (IL-27/IL-35) (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Epstein-Barr virus induced gene 3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: No test available; ED₅₀ ≤NA; Bioactivity ≥NA
- Gene ID:O35228
- Reconstitution Instructions:Sterile 20 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:IL-35/EBI3|IL-27 EBI3 subunit|IL-35 EBI3 subunit
- Protein/Peptide Name:EBI3 Subunit
- Purity:≥90%
- Molecular Weight:23 kDa
- Sequence:MALVALSQPR VQCHASRYPV AVDCSWTPLQ APNSTRSTSF IATYRLGVAT QQQSQPCLQR SPQASRCTIP DVHLFSTVPY MLNVTAVHPG GASSSLLAFV AERIIKPDPP EGVRLRTAGQ RLQVLWHPPA SWPFPDIFSL KYRLRYRRRG ASHFRQVGPI EATTFTLRNS KPHAKYCIQV SAQDLTDYGK PSDWSLPGQV ESAPHKP
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT