Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant LIF (from E. coli)
Mouse Recombinant LIF (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Mouse Recombinant LIF (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: IL-6 production by M1 cells; ED₅₀ ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
  • Gene ID:
    P09056
  • Reconstitution Instructions:
    Sterile 10 mM acetic acid at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Leukocyte Inhibitory Factor
  • Protein/Peptide Name:
    LIF
  • Purity:
    ≥95%
  • Molecular Weight:
    20 kDa
  • Sequence:
    MSPLPITPVN ATCAIRHPCH GNLMNQIKNQ LAQLNGSANA LFISYYTAQG EPFPNNVEKL CAPNMTDFPS FHGNGTEKTK LVELYRMVAY LSASLTNITR DQKVLNPTAV SLQVKLNATI DVMRGLLSNV LCRLCNKYRV GHVDVPPVPD HSDKEAFQRK KLGCQLLGTY KQVISVVVQA F
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications