Mouse Recombinant RELM-A (from E. coli)
About this item
Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77752-210
- 77752-212
- 77752-214
- 77752-216
- 77752-218
- Protein/Peptide Name:
- RELM-A
- RELM-A
- RELM-A
- RELM-A
- RELM-A
- Protein Synonyms:
- FIZZ1
- FIZZ1
- FIZZ1
- FIZZ1
- FIZZ1
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q9EP95
- Q9EP95
- Q9EP95
- Q9EP95
- Q9EP95
- Sequence:
- MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- Protease-free:
- N
- N
- N
- N
- N
- Animal-Free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-Free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Biological Activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM Sodium phosphate, pH 7.5
- 10 mM Sodium phosphate, pH 7.5
- 10 mM Sodium phosphate, pH 7.5
- 10 mM Sodium phosphate, pH 7.5
- 10 mM Sodium phosphate, pH 7.5
- Molecular Weight:
- 9.6 kDa
- 9.6 kDa
- 9.6 kDa
- 9.6 kDa
- 9.6 kDa
- Reconstitution Instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage Conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping Temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77752-210Protein/Peptide NameRELM-AProtein SynonymsFIZZ1Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9EP95SequenceMDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLSProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7.5Molecular Weight9.6 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions−20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77752-212Protein/Peptide NameRELM-AProtein SynonymsFIZZ1Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9EP95SequenceMDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLSProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7.5Molecular Weight9.6 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions−20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77752-214Protein/Peptide NameRELM-AProtein SynonymsFIZZ1Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9EP95SequenceMDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLSProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7.5Molecular Weight9.6 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions−20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77752-216Protein/Peptide NameRELM-AProtein SynonymsFIZZ1Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9EP95SequenceMDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLSProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7.5Molecular Weight9.6 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions−20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77752-218Protein/Peptide NameRELM-AProtein SynonymsFIZZ1Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9EP95SequenceMDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLSProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7.5Molecular Weight9.6 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions−20 °CShipping TemperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



