Order Entry
Puerto Rico
ContactUsLinkComponent
Pig Recombinant GM-CSF (from E. coli)
Pig Recombinant GM-CSF (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Pig Recombinant GM-CSF (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Pig
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤10 ng/ml; Bioactivity ≥1.0×10⁵ units/mg
  • Gene ID:
    Q29118
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    CSF-2|Pluripoietin-α|MGI-1GM
  • Protein/Peptide Name:
    GM-CSF (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    14.5 kDa
  • Sequence:
    MAPTRPPSPV TRPWQHVDAI KEALSLLNNS NDTAAVMNET VDVVCEMFDP QEPTCVQTRL NLYKQGLRGS LTRLKSPLTL LAKHYEQHCP LTEETSCETQ SITFKSFKDS LNKFLFTIPF DCWGPVKK
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications