Human Recombinant IL-11 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 11 (IL-11) is a member of the gp130 family of cytokines. IL-11 functions to promote hematopoietic stem cell proliferation and megakaryocyte differentiation. In non-hematopoietic cell populations, IL-11 stimulates acute-phase proteins, modulates the development of immunoglobulin-producing B cells, and regulates bone turnover. IL-11 binds the IL-11Rα receptor to activate JAK downstream signaling. Human IL-11 shows activity on murine cells.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: TF-1 Cell Proliferation; ED₅₀ ≤10 ng/ml; Bioactivity ≥1.0×10⁵ units/mg
- Gene ID:P20809
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:AGIF (Adipogenesis Inhibitory Factor)|oprelvekin
- Protein/Peptide Name:IL-11 (AF)
- Purity:≥95%
- Molecular Weight:19.3 kDa
- Sequence:MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay