Human Recombinant BAFF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment. BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:No biological activity data is available at this time
- Gene ID:Q9Y275
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:TALL|BLyS|THANK
- Protein/Peptide Name:BAFF (AF)
- Purity:≥90%
- Molecular Weight:18.5 kDa
- Sequence:MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT