Human Recombinant GDF-15-D Mutant (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Growth differentiation factor 15 (GDF-15) is a member of the transforming growth factor beta (TGF-β) family and is made by the placenta and cardiovascular tissues. GDF-15 regulates inflammatory and apoptotic pathways during cellular stress and injury. GDF-15 is emerging as a biomarker of early heart disease, such that increased levels of circulating GDF-15 are associated with an increased risk of developing heart failure. The GDF-15 D variant has a Histidine to an Aspartate substitution at amino acid position 7.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: No test available; ED₅₀ ≤Currently undetermined; Bioactivity ≥Currently undetermined
- Gene ID:Q99988
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:MIC-1|Placental TGF-β|Prostate differentiation factor
- Protein/Peptide Name:GDF-15-D Mutant
- Purity:≥95%
- Molecular Weight:12.4/24.8 kDa
- Sequence:MARNGDDCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLI QKTDTGVSLQ TYDDLLAKDC HCI
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT